Anti-UBA2 Goat Polyclonal Antibody
EVBIEB06098
- Antibody type:Primary
- Antigen name:ubiquitin-like modifier activating enzyme 2
- Antigen symbol:UBA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: UBA2 (ubiquitin-like modifier activating enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse