Anti-ING2 Goat Polyclonal Antibody
EVBIEB05452
- Antibody type:Primary
- Antigen name:Inhibitor of Growth Family, Member 2
- Antigen symbol:ING2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: ING2 (inhibitor of growth family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human