Anti-SEPT3 Goat Polyclonal Antibody
Catalog # EVBIEB05413
Supplier: EVEREST BIOTECH
Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase
- Antigen symbol:MARCH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Antigen synonyms:AXOT|RNF177|AXO|MARCH-VII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SEPT3 (septin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: