Anti-CYTIP Goat Polyclonal Antibody
EVBIEB05366
- Antibody type:Primary
- Antigen name:cytohesin 1 interacting Protein
- Antigen symbol:CYTIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Dog
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CYTIP (cytohesin 1 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Pig