Anti-ERBB2 Rabbit Polyclonal Antibody
Supplier: NOVUS BIOLOGICALS
Certificates
About this item
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP2-29624SS
- NOVUNBP2-29624
- Antigen symbol:
- ERBB2
- ERBB2
- Antigen name:
- Erb-b2 receptor tyrosine kinase 2
- Erb-b2 receptor tyrosine kinase 2
- Antigen synonyms:
- NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2064
- 2064
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together