Anti-WFDC2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27299
Supplier: NOVUS BIOLOGICALS
Specifications
- Antibody type:Primary
- Antigen name:WAP four-disulfide core domain 2
- Antigen symbol:WFDC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:10406
- Antigen synonyms:EDDM4|WAP5|dJ461P17.6|HE4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: WFDC2 (WAP four-disulfide core domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human