Order Entry
Slovakia
ContactUsLinkComponent
Anti-LXN Rabbit Polyclonal Antibody
Anti-LXN Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24530
Supplier:  NOVUS BIOLOGICALS
undefined
Anti-LXN Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24530
Supplier:  NOVUS BIOLOGICALS

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    latexin
  • Antigen symbol:
    LXN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    56925
  • Antigen synonyms:
    ECI|TCI
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: LXN (latexin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat