Anti-CNOT4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24527
Supplier: NOVUS BIOLOGICALS
Specifications
- Antibody type:Primary
- Antigen name:CCR4-NOT transcription complex, subunit 4
- Antigen symbol:CNOT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:4850
- Antigen synonyms:NOT4H|CLONE243|NOT4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: CNOT4 (CCR4-NOT transcription complex, subunit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: