Order Entry
Slovakia
ContactUsLinkComponent
Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 650) [clone: HIC1-5F10]
Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 650) [clone: HIC1-5F10]
Catalog # NOVUNBP1-18948C
Supplier:  NOVUS BIOLOGICALS
Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 650) [clone: HIC1-5F10]
Catalog # NOVUNBP1-18948C
Supplier:  NOVUS BIOLOGICALS

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heterochromatin Protein 4
  • Antigen symbol:
    HPI4
  • Clonality:
    Monoclonal
  • Clone:
    HIC1-5F10
  • Conjugation:
    DyLight® 650
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: HPi4
Clonality: monoclonal
Clone: HIC1-5F10
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human