Order Entry
Slovakia
ContactUsLinkComponent
Anti-NCL Rabbit Polyclonal Antibody
Anti-NCL Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS
Certificates
undefined
Anti-NCL Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS

Specifications

  • Catalog No:
  • NOVUNB600-241
  • NOVUNB600-241SS
  • Antigen symbol:
  • NCL
  • NCL
  • Antigen name:
  • Nucleolin
  • Nucleolin
  • Antigen synonyms:
  • Nucleolin|FLJ45706|C23|NCL|nucleolin
  • Nucleolin|FLJ45706|C23|NCL|nucleolin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4691
  • 4691
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together