Order Entry
Slovakia
ContactUsLinkComponent
Anti-CALR Rabbit Polyclonal Antibody
Anti-CALR Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS
Certificates
Anti-CALR Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS

Specifications

  • Catalog No:
  • NOVUNB600-101
  • NOVUNB600-101SS
  • Antigen symbol:
  • CRP55
  • CRP55
  • Antigen name:
  • Calreticulin
  • Calreticulin
  • Antigen synonyms:
  • CRT|FLJ26680|CALR|cC1qR|SSA|endoplasmic reticulum resident protein 60|HEL-S-99n|RO
  • CRT|FLJ26680|CALR|cC1qR|SSA|endoplasmic reticulum resident protein 60|HEL-S-99n|RO
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 811
  • 811
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together