Order Entry
Slovakia
ContactUsLinkComponent
Anti-TRPM7 Rabbit Polyclonal Antibody
Anti-TRPM7 Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS
Certificates
undefined
Anti-TRPM7 Rabbit Polyclonal Antibody
Supplier:  NOVUS BIOLOGICALS

Specifications

  • Catalog No:
  • NOVUNB500-243
  • NOVUNB500-243SS
  • Antigen symbol:
  • TRPM7
  • TRPM7
  • Antigen name:
  • Transient Receptor Potential Cation Channel Subfamily M member 7
  • Transient Receptor Potential Cation Channel Subfamily M member 7
  • Antigen synonyms:
  • TRPPLIK|CHAK1|LTrpC-7|2310022G15Rik|Ltrpc7|Ltpr7|CHAK|4833414K03Rik|5033407O22Rik
  • TRPPLIK|CHAK1|LTrpC-7|2310022G15Rik|Ltrpc7|Ltpr7|CHAK|4833414K03Rik|5033407O22Rik
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 54822
  • 54822
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together