Anti-PCNA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: PC10]
NOVUNB500-106PE
- Antibody type:Primary
- Antigen name:Proliferating cell nuclear antigen
- Antigen symbol:PCNA
- Clonality:Monoclonal
- Clone:PC10
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Gene ID:5111
- Antigen synonyms:ATLD2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone: PC10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: