Anti-HIP1 Mouse Monoclonal Antibody [clone: 4B10]
Supplier: NOVUS BIOLOGICALS
Certificates
About this item
Type: Primary
Antigen: HIP1 (huntingtin interacting protein 1)
Clonality: monoclonal
Clone: 4B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNB300-203
- NOVUNB300-203SS
- Antigen symbol:
- HIP1
- HIP1
- Antigen name:
- huntingtin interacting protein 1
- huntingtin interacting protein 1
- Antigen synonyms:
- HIP-I|ILWEQ|SHONgamma|SHONbeta|SHON
- HIP-I|ILWEQ|SHONgamma|SHONbeta|SHON
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4B10
- 4B10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3092
- 3092
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together