Anti-Sodium Potassium ATPase Alpha 1 Mouse Monoclonal Antibody [clone: 464.6]
Supplier: NOVUS BIOLOGICALS
Certificates
About this item
Type: Primary
Antigen: Sodium Potassium ATPase Alpha 1
Clonality: monoclonal
Clone: 464.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- NOVUNB300-146
- NOVUNB300-146SS
- Antigen symbol:
- ATP1A1
- ATP1A1
- Antigen name:
- Sodium Potassium ATPase Alpha 1
- Sodium Potassium ATPase Alpha 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 464.6
- 464.6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 476
- 476
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together