Anti-HIST3H3 Rabbit Polyclonal Antibody
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- NOVUNB21-1092SS
- NOVUNB21-1092
- Antigen symbol:
- H3
- H3
- Antigen name:
- Histone H3
- Histone H3
- Antigen synonyms:
- H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|H3T|HIST3H3|H3FH|MGC126886|H3FD|H3FF|HIST1H3H|H3FC|H3FB|H3FT|H3FL|H3FK|HIST1H3A
- H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|H3T|HIST3H3|H3FH|MGC126886|H3FD|H3FF|HIST1H3H|H3FC|H3FB|H3FT|H3FL|H3FK|HIST1H3A
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 126961
- 126961
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together