Anti-NTHL1 Rabbit Polyclonal Antibody
Supplier: NOVUS BIOLOGICALS
Certificates
About this item
Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-302SS
- NOVUNB100-302
- Antigen symbol:
- NTH1
- NTH1
- Antigen name:
- Endonuclease III like 1
- Endonuclease III like 1
- Antigen synonyms:
- OCTS 3|Endonuclease III like 1|NTHL 1
- OCTS 3|Endonuclease III like 1|NTHL 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 4913
- 4913
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together