Anti-Prion protein PrP Mouse Monoclonal Antibody [clone: F89/160.1.5]
Catalog # ABCAAB2777-200
Supplier: ABCAM
Specifications
- Antibody type:Primary
- Antigen name:Prion Protein PrP
- Clonality:Monoclonal
- Clone:F89/160.1.5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:1358
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pack type:Vial
- Pk:200 µG
Specifications
About this item
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Prion protein PrP
Clonality: monoclonal
Clone: F89/160.1.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: