Anti-Verotoxin II (SLT-2a, STX-2a) Mouse Monoclonal Antibody
BIRBORB23859-100
: BIORBYT
- Antibody type:Primary
- Antigen name:Verotoxin II
- Antigen symbol:SLT-2a
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:STX-2a
- Storage buffer:0,01M PBS, pH 7,2This product contains no stabilizing proteins.
- Storage temperature:Cool
- Concentration:1 mg/ml (OD280, E(0,1%)=1,3)
- Shipping temperature:Ice
- Immunogen:Purified toxoid
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Verotoxin II (SLT-2a, STX-2a)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: