Anti-PARP16 Mouse Monoclonal Antibody [clone: OTI4E2]
ORIGTA500453
- Antibody type:Primary
- Antigen name:Poly (ADP-ribose) polymerase 16
- Antigen symbol:PARP16
- Clonality:Monoclonal
- Clone:OTI4E2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:36.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Flow Cytometry: 1:100
Type: Primary
Antigen: PARP16 (poly (ADP-ribose) polymerase family, member 16)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human