Anti-WNT8B Rabbit Polyclonal Antibody
Catalog # ORIGTA317526
Supplier: ORIGENE TECHNOLOGIES
Specifications
- Antibody type:Primary
- Antigen name:wingless-type MMTV integration site family, member 8B
- Antigen symbol:WNT8B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Specifications
About this item
Recommended Dilutions: Immunochemistry: 10 µg/ml
Type: Primary
Antigen: WNT8B (wingless-type MMTV integration site family, member 8B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse