Order Entry
Slovakia
ContactUsLinkComponent
Anti-WNT8A Rabbit Polyclonal Antibody
Anti-WNT8A Rabbit Polyclonal Antibody
  ORIGTA317524
Anti-WNT8A Rabbit Polyclonal Antibody
  ORIGTA317524

 

  • Antibody type:
    Primary
  • Antigen name:
    wingless-type MMTV integration site family, member 8A
  • Antigen symbol:
    WNT8A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Pk:
    50 µG

 

 

Recommended Dilutions: Immunochemistry: 10-20 µg/ml

Type: Primary
Antigen: WNT8A (wingless-type MMTV integration site family, member 8A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human