Anti-PKD1 Goat Polyclonal Antibody
ORIGTA317165
- Antibody type:Primary
- Antigen name:Polycystic kidney disease 1
- Antigen symbol:PKD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:C-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Recommended Dilutions: Immunochemistry: 3.75 µg/ml
Type: Primary
Antigen: PKD1 (polycystic kidney disease 1 (autosomal dominant))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human