Anti-NEK11L Rabbit Polyclonal Antibody
Catalog # ABGEAP8070B
Supplier: ABGENT
Specifications
- Antibody type:Primary
- Antigen name:Stanniocalcin 2
- Antigen symbol:STC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal (450–550 aa)
- Cross adsorption:No
- Gene ID:8614
- Antigen synonyms:stanniocalcin-related protein|STCRP|stanniocalcin-2|STC-2|STC-related protein
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:CSEWEDEQSEYSDIRR
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This STC2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 273-302 amino acids from the C-terminal region of human STC2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NEK11L
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse