Anti-RIPK4 Rabbit Polyclonal Antibody
ABGEAP7808B
: ABGENT
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 4
- Antigen symbol:RIPK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal (800–900 aa)
- Cross adsorption:No
- Antigen synonyms:Ankyrin repeat domain protein 3|PKC-delta-interacting protein kinase|Serine/threonine-protein kinase ANKRD3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: RIPK4 (receptor-interacting serine-threonine kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse