Order Entry
Slovakia
ContactUsLinkComponent
Anti-MMP8 Rabbit Polyclonal Antibody
Anti-MMP8 Rabbit Polyclonal Antibody
Catalog # ABGEAP6213A
Supplier:  ABGENT
undefined
Anti-MMP8 Rabbit Polyclonal Antibody
Catalog # ABGEAP6213A
Supplier:  ABGENT

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 8
  • Antigen symbol:
    MMP8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Center
  • Cross adsorption:
    No
  • Antigen synonyms:
    ADAMTS19
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MMP8 (matrix metallopeptidase 8 (neutrophil collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse