Anti-BBS10 Rabbit Polyclonal Antibody
ABGEAP4880B
: ABGENT
- Antibody type:Primary
- Antigen name:Bardet-Biedl Syndrome 10
- Antigen symbol:BBS10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:Bardet Biedl syndrome 10 protein|FLJ23560|BBS10_HUMAN.|C12orf58|Bardet Biedl syndrome 10 protein homolog|RGD1560748
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: BBS10 (Bardet-Biedl syndrome 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human