Order Entry
Slovakia
ContactUsLinkComponent
Anti-NSE Rabbit Polyclonal Antibody
Anti-NSE Rabbit Polyclonal Antibody
  ABGEAP2780A
 :  ABGENT
undefined
Anti-NSE Rabbit Polyclonal Antibody
  ABGEAP2780A
 :  ABGENT

 

  • Antibody type:
    Primary
  • Antigen name:
    Enolase 2 (gamma, neuronal)
  • Antigen symbol:
    ENO2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    Gamma-enolase|Enolase 2|ENO2|Neural enolase|ENO-2|2-phospho-D-glycerate hydro-lyase|Neuron-specific enolase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: NSE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human