Anti-FBL Rabbit Polyclonal Antibody
Catalog # ABGEAP2776C
Supplier: ABGENT
Specifications
- Antibody type:Primary
- Antigen name:Fibrillarin
- Antigen symbol:FBL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:FIB|FLRN|RNU3IP1
- Sequence:E120
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human