Order Entry
Slovakia
ContactUsLinkComponent
Anti-UBE2V1 Rabbit Polyclonal Antibody
Anti-UBE2V1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2157B
Supplier:  ABGENT
undefined
Anti-UBE2V1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2157B
Supplier:  ABGENT

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin-conjugating Enzyme E2 variant 1
  • Antigen symbol:
    UBE2V1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    UEV1|Ubiquitin-conjugating enzyme E2 variant 1|UEV1A|UBE2V|P/OKcl.19|CROC-1|CIR1|UEV-1|CROC1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: UBE2V1 (ubiquitin-conjugating enzyme E2 variant 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse