Anti-USP14 Rabbit Polyclonal Antibody
ABGEAP2142A
: ABGENT
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 14
- Antigen symbol:USP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal ( 1–50 aa)
- Cross adsorption:No
- Antigen synonyms:ENK|ecat4|2410002E02Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: USP14 (ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human