Anti-USP12/USP46 Rabbit Polyclonal Antibody
Catalog # ABGEAP2140B
Supplier: ABGENT
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 12
- Antigen symbol:USP12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: USP12/USP46
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human