Anti-MOB4 Rabbit Polyclonal Antibody
Catalog # ABGEAP16890A
Supplier: ABGENT
Specifications
- Antibody type:Primary
- Antigen name:MOB family member 4, phocein
- Antigen symbol:MOB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:25843
- Antigen synonyms:2C4D|MOB1|MOBKL3|PHOCN|MOB3|PREI3|CGI-95
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:26.032 kDa
- Sequence:TAVLRRNRPGTKAQDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MOBKL3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human MOBKL3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MOB4 (MOB family member 4, phocein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human