Anti-TLR3 Rabbit Polyclonal Antibody
ABGEAP1503E
: ABGENT
- Antibody type:Primary
- Antigen name:Toll Like Receptor 3
- Antigen symbol:TLR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:IIAE2|CD283|TLR3|Toll-like receptor 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human