Anti-NR2F2 Rabbit Polyclonal Antibody
ABGEAP14204A
: ABGENT
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 2, Group F, Member 2
- Antigen symbol:NR2F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:7026
- Antigen synonyms:ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:45.571 kDa
- Sequence:RDPQDEVPGSQGSQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NR2F2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human NR2F2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: NR2F2 (nuclear receptor subfamily 2, group F, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human