Anti-CIR1 Rabbit Polyclonal Antibody
ABGEAP1417A
: ABGENT
- Antibody type:Primary
- Antigen name:Corepressor Interacting with RBPJ, 1
- Antigen symbol:CIR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CIR
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CIR1 (corepressor interacting with RBPJ, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human