Anti-ANKRD6 Rabbit Polyclonal Antibody
ABGEAP13033C
: ABGENT
- Antibody type:Primary
- Antigen name:Ankyrin repeat domain 6
- Antigen symbol:ANKRD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:ANKRD6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ANKRD6 (ankyrin repeat domain 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human