Anti-EAPP Rabbit Polyclonal Antibody
ABGEAP12971B
: ABGENT
- Antibody type:Primary
- Antigen name:E2F-associated phosphoprotein
- Antigen symbol:EAPP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: EAPP (E2F-associated phosphoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human