Anti-MORN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP12954C
Supplier: ABGENT
Specifications
- Antibody type:Primary
- Antigen name:Catenin Delta 1
- Antigen symbol:MORN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:CTNND|p120ctn|P120CAS|p120(ctn)|p120|p120(cas)|P120CTN|CAS |KIAA0384|p120cas
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MORN3 (MORN repeat containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human