Anti-YY1 Rabbit Polyclonal Antibody
AVIVP100899_P050
- Antibody type:Primary
- Antigen name:YY1 transcription factor
- Antigen symbol:YY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7528
- Antigen synonyms:YIN-YANG-1|UCRBP|INO80S|DELTA|NF-E1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: YY1 (YY1 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat