Anti-DBF4B Rabbit Polyclonal Antibody
Catalog # AVIVARP70104_P050
Supplier: AVIVA SYSTEMS BIOLOGY
Specifications
- Antibody type:Primary
- Antigen name:DBF4 zinc finger B
- Antigen symbol:DBF4B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:80174
- Antigen synonyms:ASKL1, CHIFB, DRF1, ZDBF1B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DBF4B (DBF4 zinc finger B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human