Deuterium lamps
Catalog # HEWL2140-0590
Supplier: AGILENT HEWLETT PACKARD
Specifications
- For:G1315A/B, G1365A/B
- Description:Deuterium lamp
- Seq:01
- Pk:1
Specifications
About this item
These deuterium lamps are designed with a much narrower aperture providing increased light intensity and decreased noise, and tested for noise and drift specifications, correct operating voltage, light intensity and proper alignment.
- Improved coating
- Increasing lifetime to up to 2000 hours of use
- Highly sensitive lamps, extends detection at trace levels
Certifications: Manufactured in ISO 9001 environment and fully traceable, calibrated using optical standards provided by NIST, PTB.