Chicken blood, sterile
Catalog # ROCKR302-0050
Supplier: ROCKLAND
Specifications
- Description:Chicken blood in Alsever's
- Seq:01
- Pk:50 mL
Specifications
About this item
Chicken whole blood aseptically collected. These cell suspensions are ideal for washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.