Iron and manganese test kit, colour disc, IR-20
Catalog # HACH146300
Supplier: HACH
CAS Number:
Specifications
- Pk:1 KIT
- Description:Iron and manganese colour disc test kit, IR-20
- Range:0 - 4 mg/l Fe
Specifications
About this item
Test kit suitable to detect the presence of iron and manganese in cases of stained laundry.
- Simple and fast colour comparison method
- Portable test kit for on site measurement
Delivery: Includes three reagents, bottles, test tube brush, colour discs, comparator box, viewing adapter, viewing tubes, instructions and carrying case.