Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Rotameters, Sigma-Aldrich
Rotameters, Sigma-Aldrich
Supplier:   Merck
Rotameters, Sigma-Aldrich
Supplier:  Merck
Certificates
Rotameters, Sigma-Aldrich
3 Options Available Below
Product Details & Documents
These rotameters are designed for accurate metering and chromatographic purposes.
  • Flow tube and four interchangeable floats
  • Standard flow tables (ml/minute at STP) included
  • Can be mounted or installed on the (optional) tripod assembly
Suitable for chromatography applications.
Product Family Options
Needle valve
Product Information
  • Flow rate
    Accuracy
    Connector
    Pressure
    Length
    Availability
    Price
  • 0 - 33 ml/min
    ±2% full scale
    1/8” female NPT
    13,8 bar
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 280,31 €
    280,31 €
    1 * 1 Each
       280,31 €
  • 0 - 246 ml/min
    ±2% full scale
    1/8” female NPT
    13,8 bar
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 441,84 €
    441,84 €
    1 * 1 Each
       441,84 €
  • 0 - 454 ml/min
    ±2% full scale
    1/8” female NPT
    13,8 bar
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 413,89 €
    413,89 €
    1 * 1 Each
       413,89 €