Washing solution, Protein-OUT™
Catalog # 786-680
Supplier: G BIOSCIENCES
CAS Number:
Specifications
- Pk:50 mL
- Description:ProteinOUT™ washing solution
Specifications
About this item
Protein-OUT™ is a washing solution that cleans plastic and glassware used in the proteolytic digestion and peptide isolation for mass spectrometry analysis. The rinsing of plastic and glassware with Protein-OUT™ will remove interfering chemicals and peptides that can leach from the tube walls and interfere or suppress the mass spectrometry signal. In addition, rinsing with Protein-OUT™ removes inadvertent contamination with dust and keratin.
- Cost effective alternative to siliconised tubes
- Supplied in 50 ml bottles
- Suitable for the rinsing of ~200 1,5 - 2 ml micro centrifuge tubes