ESD glove liners, A198
Supplier: Portwest
Certificates
About this item
These liners feature a nylon and carbon fibre shell which diverts static electricity.
- Fingertips with PU coating for precision applications
- Excellent dexterity and durability
For use in electronics assembly, testing and precision work.
Colour: Grey/white
EN 388 (2132), EN 1149
Specifications
- Catalog No:
- PRTWA198GRRXL
- PRTWA198GRRL
- PRTWA198GRRM
- PRTWA198GRRS
- PRTWA198GRRXS
- Model:
- A198
- A198
- A198
- A198
- A198
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Gauge:
- 13
- 13
- 13
- 13
- 13
- EN norm:
- EN 388 (2132), EN 1149
- EN 388 (2132), EN 1149
- EN 388 (2132), EN 1149
- EN 388 (2132), EN 1149
- EN 388 (2132), EN 1149
- Anti-static:
- +
- +
- +
- +
- +