Order Entry
Slovakia
ContactUsLinkComponent
Cold resistant gloves, Arctic Winter Glove, A146
Cold resistant gloves, Arctic Winter Glove, A146
Supplier:  Portwest
Certificates
undefined
Cold resistant gloves, Arctic Winter Glove, A146
Supplier:  Portwest
Specifications
  • Catalog No:
  • PRTWA146YERXL
  • PRTWA146YERL
  • PRTWA146YERXXL
  • Model:
  • Arctic Winter, A146
  • Arctic Winter, A146
  • Arctic Winter, A146
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon
  • Nylon
  • Nylon
  • Finish:
  • 3/4 micro foam nitrile dipping
  • 3/4 micro foam nitrile dipping
  • 3/4 micro foam nitrile dipping
  • Gauge:
  • 15
  • 15
  • 15
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4242X), EN 407 (X2XXXX), EN 511 (X2X)
  • EN 388:2016 (4242X), EN 407 (X2XXXX), EN 511 (X2X)
  • EN 388:2016 (4242X), EN 407 (X2XXXX), EN 511 (X2X)