Protective hand foam, Schaum-A-Derm
Catalog # 115-3249
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Specifications
- Description:Protective hand foam, Schaum-A-Derm, for laboratory, 150 ml
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The skin protection foam protects the skin over a period of four hours against substances such as cleaning agents and detergents, organic solvents, acids and caustics, paints and varnishes. Rub a walnut-sized amount onto dry, clean hands and allow to absorb for only two minutes. The unit provides approx. 150 applications.
- Maintains the skin's sense of touch and ability to grip
- Forms a thin, viscoelastic, non-stick film on the skin
- Skin continues to breathe and transpire naturally
- Dermatologically harmless, preferred by allergy sufferers
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904