2-Deoxy-D-glucose
Supplier: CAYMAN
Jump to:
About this item
Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal
- CAS Number:154-17-6
- Formula:C₆H₁₂O₅
- MDL Number:MFCD00151328
- Molecular Weight:164.16 g/mol
- EINECS:205-823-0
4 Options Available Below
Product Details & Documents
Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal
- CAS Number:154-17-6
- Formula:C₆H₁₂O₅
- MDL Number:MFCD00151328
- Molecular Weight:164.16 g/mol
- EINECS:205-823-0
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



